5-Chloro-2-fluoro-3-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Danger

A112266-1G A112266-250MG
76765-928EA 159 USD
76765-928 76765-930
5-Chloro-2-fluoro-3-(trifluoromethyl)pyridine 95%
5-Chloro-2-fluoro-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₄N
MW: 199.53 g/mol
MDL Number: MFCD09749652
CAS Number: 71701-96-7
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR