6-Bromoimidazo[1,2-a]pyrimidine 97%

Supplier: Ambeed
Warning

A112205-10G A112205-1G A112205-250MG A112205-25G A112205-5G
76765-918EA 162 USD
76765-918 76765-920 76765-922 76765-924 76765-926
6-Bromoimidazo[1,2-a]pyrimidine 97%
6-Bromoimidazo[1,2-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MDL Number: MFCD09261434
CAS Number: 865156-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR