5-Bromo-3-fluoro-2-pyridine carboxylic acid 98%

Supplier: Ambeed
Warning

A111645-10G A111645-1G A111645-250MG A111645-5G A111645-25G
76765-868EA 113 USD
76765-868 76765-870 76765-872 76765-874 77595-746
5-Bromo-3-fluoro-2-pyridine carboxylic acid 98%
5-Bromo-3-fluoro-2-pyridine carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MDL Number: MFCD07437865
CAS Number: 669066-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR