6-Chloro-2-hydroxymethylpyridine hydrochloride 95%

Supplier: Ambeed
Warning

A111368-1G A111368-5G
76765-834EA 46 USD
76765-834 76765-836 76765-838
6-Chloro-2-hydroxymethylpyridine hydrochloride 95%
6-Chloro-2-hydroxymethylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₂NO
MDL Number: MFCD08690070
CAS Number: 83782-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR