5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A111218-1G A111218-250MG A111218-25G A111218-5G
76765-822EA 50.93 USD
76765-822 76765-824 76765-826 76765-828
5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid 95%
5-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂O₂
MW: 222.6300 g/mol
MDL Number: MFCD03715908
CAS Number: 54006-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR