2-Amino-3-(6-chloro-1H-indol-3-yl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-DL-Trp(6-Cl)-OH

A110000-1G A110000-250MG
76765-754EA 97 USD
76765-754 76765-756
2-Amino-3-(6-chloro-1H-indol-3-yl)propanoic acid 95%
2-Amino-3-(6-chloro-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂O₂
MW: 238.67 g/mol
MDL Number: MFCD00152036
CAS Number: 17808-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR