2-Fluoro-5-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A109986-1000G A109986-100G A109986-10G A109986-25G A109986-500G A109986-5G
76765-742EA 482 USD
76765-742 76765-744 76765-746 76765-748 76765-750 76765-752
2-Fluoro-5-nitrobenzonitrile 98%
2-Fluoro-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FN₂O₂
MDL Number: MFCD00042299
CAS Number: 17417-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR