STAT3 Inhibitor V, Stattic 98%

Supplier: Ambeed
Warning

A109795-10MG A109795-1MG A109795-25MG A109795-50MG A109795-100MG A109795-250MG
76765-728EA 13 USD
76765-728 76765-730 76765-732 76765-734 77599-580 77623-910
STAT3 Inhibitor V, Stattic 98%
STAT3 Inhibitor V, Stattic

Precaución: for Research Use Only

Formula: C₈H₅NO₄S
MDL Number: MFCD00173799
CAS Number: 19983-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR