4-(Boc-amino)cyclohexanone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Aminocyclohexan-1-one, N-BOC protected, tert-Butyl (4-oxocyclohexyl)carbamate

A109484-100G A109484-10G A109484-1G A109484-25G A109484-500G A109484-5G
76765-708EA 80 USD
76765-708 76765-710 76765-712 76765-714 76765-716 76765-718
4-(Boc-amino)cyclohexanone 97%
4-(Boc-amino)cyclohexanone

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.2700 g/mol
MDL Number: MFCD00798168
CAS Number: 179321-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR