Ethionamide 98%

Supplier: Ambeed
Warning

A109412-10G A109412-1G A109412-25G A109412-5G A109412-100G
76765-700EA 30 USD
76765-700 76765-702 76765-704 76765-706 77623-904
Ethionamide 98%
Ethionamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂S
MDL Number: MFCD00057361
CAS Number: 536-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR