4-tert-Butylcyclohexylamine 95%

Supplier: Ambeed
Danger

Synonyms: 4-(tert-Butyl)cyclohexanamine

A108832-1G A108832-250MG A108832-25G A108832-5G
76765-662EA 21.22 USD
76765-662 76765-664 76765-666 77570-506
4-tert-Butylcyclohexylamine 95%
4-tert-Butylcyclohexylamine

Precaución: for Research Use Only

Formula: C₁₀H₂₁N
MW: 155.28 g/mol
MDL Number: MFCD00013763
CAS Number: 5400-88-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR