2,3,5-Trimethylanisole 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methoxy-2,3,5-trimethylbenzene

A108655-100G A108655-25G A108655-5G
76765-656EA 282.2 USD
76765-656 76765-658 76765-660
2,3,5-Trimethylanisole 95%
2,3,5-Trimethylanisole

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD00142783
CAS Number: 20469-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR