5-(2,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A108637-1G A108637-250MG A108637-5G
76765-650EA 71.09 USD
76765-650 76765-652 76765-654
5-(2,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid 95%
5-(2,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆Cl₂N₂O₂
MW: 257.0700 g/mol
MDL Number: MFCD04969776
CAS Number: 78874-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR