6-Chloro-5-nitropyrimidine-2,4-diol 97%

Supplier: Ambeed
Warning

A108528-100MG A108528-1G A108528-250MG
76765-644EA 53 USD
76765-644 76765-646 76765-648
6-Chloro-5-nitropyrimidine-2,4-diol 97%
6-Chloro-5-nitropyrimidine-2,4-diol

Precaución: for Research Use Only

Formula: C₄H₂ClN₃O₄
MDL Number: MFCD09753676
CAS Number: 6630-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR