Methyl-5-phenylisoxazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-phenylisoxazole-3-carboxylate

A107984-10G A107984-5G
76765-606EA 254.62 USD
76765-606 76765-608
Methyl-5-phenylisoxazole-3-carboxylate 95%
Methyl-5-phenylisoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.1941 g/mol
MDL Number: MFCD02159553
CAS Number: 51677-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR