5-Amino-3-phenylisoxazole 97%

Supplier: Ambeed
Warning

A107659-1G A107659-250MG A107659-5G
76765-600EA 42 USD
76765-600 76765-602 76765-604
5-Amino-3-phenylisoxazole 97%
5-Amino-3-phenylisoxazole

Precaución: for Research Use Only

Formula: C₉H₇NO
MDL Number: MFCD00464114
CAS Number: 4369-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR