1-Phenyl-2-urea 98%

Supplier: Ambeed
Warning

Synonyms: Phenylurea, 1-Phenylurea, N-Phenylurea

A107537-100G A107537-25G A107537-500G
76765-594EA 8.49 USD
76765-594 76765-596 76765-598
1-Phenyl-2-urea 98%
1-Phenyl-2-urea

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD00007944
CAS Number: 64-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR