5-(3-Chlorophenyl)-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A107416-10G A107416-1G A107416-250MG A107416-25G A107416-5G
76765-576EA 279.02 USD
76765-576 76765-578 76765-580 76765-582 76765-584
5-(3-Chlorophenyl)-1H-pyrazole-3-carboxylic acid 95%
5-(3-Chlorophenyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂O₂
MW: 222.6300 g/mol
MDL Number: MFCD05170019
CAS Number: 595610-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR