3',5'-Bis(trifluoromethyl)acetophenone 98%

Supplier: Ambeed
Warning

A107409-10G A107409-25G A107409-5G A107409-100G A107409-500G
76765-570EA 6 USD
76765-570 76765-572 76765-574 77549-070 77570-450
3',5'-Bis(trifluoromethyl)acetophenone 98%
3',5'-Bis(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₀H₆F₆O
MDL Number: MFCD00009910
CAS Number: 30071-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR