2,4-Dihydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A107043-10G A107043-1G A107043-250MG A107043-25G A107043-5G
76765-530EA 98 USD
76765-530 76765-532 76765-534 76765-536 77876-814
2,4-Dihydroxybenzonitrile 98%
2,4-Dihydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅NO₂
MDL Number: MFCD02261931
CAS Number: 64419-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR