2,6-Difluoro-4-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A106845-100MG A106845-1G A106845-250MG A106845-25G A106845-5G
76765-520EA 15.92 USD
76765-520 76765-522 76765-524 76765-526 76765-528
2,6-Difluoro-4-hydroxybenzoic acid 95%
2,6-Difluoro-4-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄F₂O₃
MDL Number: MFCD03094500
CAS Number: 214917-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR