Boc-D-valinol 95%

Supplier: Ambeed
Warning

Synonyms: N-BOC-D-Valinol, Boc-D-Valinol

A106821-100G A106821-10G A106821-1G A106821-25G A106821-5G
76765-510EA 195.21 USD
76765-510 76765-512 76765-514 76765-516 76765-518
Boc-D-valinol 95%
Boc-D-valinol

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
MDL Number: MFCD00235960
CAS Number: 106391-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR