6-Chloroimidazo[1,2-a]pyridine-8-carboxylic acid 97%

Supplier: Ambeed
Warning

A106586-10G A106586-1G A106586-5G A106586-100MG A106586-250MG
76765-498EA 245 USD
76765-498 76765-500 76765-502 77570-392 77876-813
6-Chloroimidazo[1,2-a]pyridine-8-carboxylic acid 97%
6-Chloroimidazo[1,2-a]pyridine-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD09031676
CAS Number: 155735-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR