6-Chloroanthranilic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-chlorobenzoic acid

A106388-100G A106388-10G A106388-1G A106388-25G A106388-500G
76765-486EA 32.89 USD
76765-486 76765-488 76765-490 76765-492 76765-494
6-Chloroanthranilic acid 97%
6-Chloroanthranilic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD00051530
CAS Number: 2148-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR