6-Amino-1,3-benzoxazol-2(3H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2-hydroxybenzoxazole, 6-Amino-2-benzoxazolone, 6-Amino-2-benzoxazolinone

A105475-100MG A105475-1G A105475-250MG A105475-5G
76765-464EA 19.1 USD
76765-464 76765-466 76765-468 76765-470
6-Amino-1,3-benzoxazol-2(3H)-one 95%
6-Amino-1,3-benzoxazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₆N₂O₂
MW: 150.14 g/mol
MDL Number: MFCD00463899
CAS Number: 22876-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR