5-Methyl-1H-indole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A105309-100MG A105309-10G A105309-250MG A105309-25G A105309-5G A105309-1G
76765-452EA 5.31 USD
76765-452 76765-454 76765-456 76765-458 76765-460 77565-838
5-Methyl-1H-indole-2-carboxylic acid 98%
5-Methyl-1H-indole-2-carboxylic acid

Precaución: For research use only.

Formula: C10H9NO2
MW: 175.19 g/mol
MDL Number: MFCD00047166
CAS Number: 10241-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR