3-(4-Morpholinyl)aniline 98%

Supplier: Ambeed
Warning

A104719-100G A104719-10G A104719-1G A104719-250MG A104719-25G A104719-5G
76765-416EA 649.28 USD
76765-416 76765-418 76765-420 76765-422 76765-424 76765-426
3-(4-Morpholinyl)aniline 98%
3-(4-Morpholinyl)aniline

Precaución: for Research Use Only

Formula: C10H14N2O
MW: 178.2300 g/mol
MDL Number: MFCD03197165
CAS Number: 159724-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR