3-(Trifluoromethoxy)benzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A104710-10G
A104710-1G
A104710-25G
A104710-5G
A104710-100G
76765-408EA
22.28
USD
76765-408
76765-410
76765-412
76765-414
77599-532
3-(Trifluoromethoxy)benzaldehyde 97%
3-(Trifluoromethoxy)benzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₅F₃O₂ MW: 190.12 g/mol |
MDL Number:
MFCD00042406 CAS Number: 52771-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...