5-Chloro-6-methylpyrimidin-4(3H)-one 95%

Supplier: Ambeed
Warning

A104634-100MG A104634-250MG A104634-1G
76765-404EA 12 USD
76765-404 76765-406 77876-802
5-Chloro-6-methylpyrimidin-4(3H)-one 95%
5-Chloro-6-methylpyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MDL Number: MFCD08234409
CAS Number: 7752-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR