(Methyl)triphenylphosphonium iodide 97%

Supplier: Ambeed
Warning

Synonyms: Methyltriphenylphosphonium iodide

A104405-10G A104405-500G A104405-5G A104405-100G A104405-25G
76765-382EA 7.43 USD
76765-382 76765-384 76765-386 77565-820 77565-822
(Methyl)triphenylphosphonium iodide 97%
(Methyl)triphenylphosphonium iodide

Precaución: For research use only.

Formula: C₁₉H₁₈IP
MW: 404.23 g/mol
MDL Number: MFCD00066175
CAS Number: 2065-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR