5-Chloro-2-(trifluoromethyl)benzyl bromide 95%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-5-chloro-2-(trifluoromethyl)benzene, 2-(Bromomethyl)-4-chloro-1-(trifluoromethyl)benzene

A104361-10G A104361-1G A104361-25G A104361-5G
76765-374EA 98.67 USD
76765-374 76765-376 76765-378 76765-380
5-Chloro-2-(trifluoromethyl)benzyl bromide 95%
5-Chloro-2-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₅BrClF₃
MW: 273.48 g/mol
MDL Number: MFCD01631356
CAS Number: 261763-24-0
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR