6-Chlorobenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A104349-100MG A104349-1G A104349-250MG
76765-368EA 21.22 USD
76765-368 76765-370 76765-372
6-Chlorobenzo[d]thiazole 97%
6-Chlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₄ClNS
MW: 169.63 g/mol
MDL Number: MFCD01325561
CAS Number: 2942-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR