4-Chloro-3-fluorobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A104035-1G
A104035-100G
A104035-10G
A104035-25G
76765-340EA
8
USD
76765-340
77570-240
77570-242
77570-244
4-Chloro-3-fluorobenzaldehyde 98%
4-Chloro-3-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄ClFO |
MDL Number:
MFCD00143288 CAS Number: 5527-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...