(S)-4-Phenyl-3-propionyloxazolidin-2-one 97%

Supplier: Ambeed
Warning

A103404-100G A103404-1G A103404-250MG A103404-25G A103404-5G
76765-320EA 366 USD
76765-320 76765-322 76765-324 76765-326 76765-328
(S)-4-Phenyl-3-propionyloxazolidin-2-one 97%
(S)-4-Phenyl-3-propionyloxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD06658223
CAS Number: 184363-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR