4-Bromo-5-fluoro-2-nitrotoluene 98%

Supplier: Ambeed
Warning

A103366-10G A103366-1G A103366-5G A103366-25G
76765-314EA 30.77 USD
76765-314 76765-316 76765-318 77623-852
4-Bromo-5-fluoro-2-nitrotoluene 98%
4-Bromo-5-fluoro-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MW: 234.02 g/mol
MDL Number: MFCD04039299
CAS Number: 224185-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR