(5-Amino-2-fluorophenyl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-fluorobenzyl alcohol

A102769-1G A102769-5G
76765-266EA 9 USD
76765-266 76765-268
(5-Amino-2-fluorophenyl)methanol 98%
(5-Amino-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.15 g/mol
MDL Number: MFCD03094257
CAS Number: 84832-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR