5-Bromo-1-methylpyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1-methyl-2(1H)-pyridinone

A102696-1G A102696-250MG A102696-25G A102696-5G A102696-10G
76765-260EA 23.34 USD
76765-260 76765-262 76765-264 77595-706 77623-840
5-Bromo-1-methylpyridin-2(1H)-one 97%
5-Bromo-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD09972188
CAS Number: 81971-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR