trans-4-Isopropylcyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

A102475-10G A102475-5G A102475-25G
76765-246EA 9.55 USD
76765-246 76765-248 77623-832
trans-4-Isopropylcyclohexanecarboxylic acid 97%
trans-4-Isopropylcyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
MDL Number: MFCD04004145
CAS Number: 7077-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR