(2,3-Dimethoxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A102473-100MG A102473-1G A102473-250MG A102473-5G
76765-238EA 15 USD
76765-238 76765-240 76765-242 76765-244
(2,3-Dimethoxyphenyl)acetic acid 98%
(2,3-Dimethoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MDL Number: MFCD00060351
CAS Number: 90-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR