5-Bromo-2-(dimethylamino)pyrimidine 98%

Supplier: Ambeed
Warning

A102377-100G A102377-10G A102377-1G A102377-250MG A102377-25G A102377-5G
76765-226EA 239 USD
76765-226 76765-228 76765-230 76765-232 76765-234 76765-236
5-Bromo-2-(dimethylamino)pyrimidine 98%
5-Bromo-2-(dimethylamino)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₈BrN₃
MDL Number: MFCD07368228
CAS Number: 38696-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR