3,6-Difluorophthalic acid 98%

Supplier: Ambeed
Warning

A102355-100MG A102355-250MG A102355-5G A102355-1G
76765-220EA 18 USD
76765-220 76765-222 76765-224 77599-498
3,6-Difluorophthalic acid 98%
3,6-Difluorophthalic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₄
MDL Number: MFCD03840499
CAS Number: 651-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR