4-(4-Fluorobenzoyl)piperidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (4-Fluorophenyl)(piperidin-4-yl)methanone hydrochloride

A102247-1G A102247-25G A102247-5G A102247-10G
76765-214EA 7.43 USD
76765-214 76765-216 76765-218 77595-704
4-(4-Fluorobenzoyl)piperidine hydrochloride 98%
4-(4-Fluorobenzoyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClFNO
MW: 243.71 g/mol
MDL Number: MFCD00044912
CAS Number: 25519-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR