3-Fluoro-o-toluidine 97%

Supplier: Ambeed
Warning

A102219-10G A102219-1G A102219-25G A102219-500G A102219-5G A102219-100G
76765-204EA 11 USD
76765-204 76765-206 76765-208 76765-210 76765-212 77876-789
3-Fluoro-o-toluidine 97%
3-Fluoro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈FN
MDL Number: MFCD00007760
CAS Number: 443-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR