3-Methyl-1,2-cyclopentanedione 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-1,2-cyclopentanedione, Maple lactone, Methyl cyclopentenolone, 3-Methylcyclopentane-1,2-dione

A102000-100G A102000-25G A102000-5G
76765-198EA 28.65 USD
76765-198 76765-200 76765-202
3-Methyl-1,2-cyclopentanedione 95%
3-Methyl-1,2-cyclopentanedione

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00001417
CAS Number: 765-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR