2-Bromo-5'-fluoro-2'-hydroxyacetophenone 98%

Supplier: Ambeed
Danger

A100260-1G A100260-250MG A100260-5G
76765-100EA 29 USD
76765-100 76765-102 76765-104
2-Bromo-5'-fluoro-2'-hydroxyacetophenone 98%
2-Bromo-5'-fluoro-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MDL Number: MFCD03094564
CAS Number: 126581-65-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR