TCM-ProteaseArrest™ Protease Inhibitor Cocktail
Supplier: G-Biosciences
786-238
786-239
76285-480EA
206.15
USD
76285-480
76285-830
TCM-ProteaseArrest™ Protease Inhibitor Cocktail
Cell Culture Supplements and Additives
TCM-ProteaseArrest™ is a mixture of protease inhibitors with a broad specificity for the inhibition of serine, cysteine, aspartic (acid) proteases and aminopeptidases.
- For tissue culture medium to prevent the degradation of secreted proteins
- Store at –20 °C
TCM-ProteaseArrest™ is supplied as a 200X concentrated solution in sterile filtered DMSO. It is highly recommended to determine the appropriate dilution factor for a specific cell line, starting at a 1:200 dilution of TCM-ProteaseArrest™ to prevent harmful effects of DMSO to cell growth.