Sodium methacrylate 98%

Supplier: Ambeed
Warning

A996470-100G A996470-10G A996470-25G A996470-500G
76729-456EA 10 USD
76729-456 76729-458 76729-460 76729-462
Sodium methacrylate 98%
Sodium methacrylate

Precaución: for Research Use Only

Formula: C₄H₅NaO₂
MW: 108.072 g/mol
MDL Number: MFCD00045886
CAS Number: 5536-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR