Potassium-4-butoxyphenyltrifluoroborate 98%

Supplier: Ambeed
Warning

A994142-1G A994142-250MG A994142-5G
76729-450EA 58 USD
76729-450 76729-452 76729-454
Potassium-4-butoxyphenyltrifluoroborate 98%
Potassium 4-butoxyphenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₁₀H₁₃BF₃KO
MW: 256.12 g/mol
MDL Number: MFCD04115755
CAS Number: 850623-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR