N-Ethylacetamide 98%

Supplier: Ambeed
Warning

A990358-100G A990358-25G A990358-5G
76729-438EA 79.57 USD
76729-438 76729-440 76729-442
N-Ethylacetamide 98%
N-Ethylacetamide

Precaución: for Research Use Only

Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD00009029
CAS Number: 625-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR