4-Acetamido-N-(2-(diethylamino)ethyl)benzamide hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: N-Acetylprocainamide HCl

A989830-100MG A989830-1G A989830-250MG
76729-432EA 15.92 USD
76729-432 76729-434 76729-436
4-Acetamido-N-(2-(diethylamino)ethyl)benzamide hydrochloride 98+%
4-Acetamido-N-(2-(diethylamino)ethyl)benzamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₄ClN₃O₂
MW: 313.82 g/mol
MDL Number: MFCD00012501
CAS Number: 34118-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR